ADV428293375 AChemBlock

(2R,4S)-4-Fluoropyrrolidine-2-carboxylic acid

  • MDL number MFCD07366527
  • Cas Number : 131176-02-8
  • Molecular Weight : 133.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2r-4s-4-fluoropyrrolidine-2-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7739
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart