ADV428293377 AChemBlock

Methyl 3-(pyrrolidine-3-yl)propanoate hydrochloride

  • MDL number MFCD11111229
  • Cas Number : 1211593-43-9
  • Molecular Weight : 193.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-pyrrolidine-3-yl-propanoate-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7741
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart