ADV428293383 AChemBlock

[(3S,4R)-rel-3-fluoro-4-piperidyl]methanol hydrochloride

  • MDL number MFCD27937749
  • Cas Number : 895577-96-5
  • Molecular Weight : 169.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3s-4r-rel-3-fluoro-4-piperidyl-methanol-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7747
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart