ADV428293387 AChemBlock

Ethyl (2R,4S)-4-fluoropyrrolidine-2-carboxylate

  • MDL number MFCD27987077
  • Cas Number : 1523530-67-7
  • Molecular Weight : 161.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2r-4s-4-fluoropyrrolidine-2-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7751
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart