ADV428293388 AChemBlock

5-Methoxy-1H-pyrazol-3-amine hydrochloride

  • MDL number MFCD17392863
  • Cas Number : 110884-53-2
  • Molecular Weight : 149.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid/white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-methoxy-1h-pyrazol-3-amine-hydrochloride.html

Safety Information

signalword none
pcodes P273;P391
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3466
pictograms GHS09
hstatements H411
Availability details for
Quantity
  • Availability
  • Add to Cart