ADV428293393 AChemBlock

2,4-Dibromo-5-nitropyridine

  • MDL number MFCD09832226
  • Cas Number : 4487-57-4
  • Molecular Weight : 281.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-4-dibromo-5-nitropyridine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7757
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart