ADV428293397 AChemBlock

(3R)-1-Boc-3-(4-amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidin-1-yl)piperidine

  • MDL number MFCD27987084
  • Cas Number : 1276110-38-3
  • Molecular Weight : 444.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3r-1-boc-3-4-amino-3-iodo-1h-pyrazolo-3-4-d-pyrimidin-1-yl-piperidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7761
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart