ADV428293398 AChemBlock

4-Amino-3-pyridinecarboxamide hydrochloride

  • MDL number MFCD18425621
  • Cas Number : 7418-66-8
  • Molecular Weight : 173.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-amino-3-pyridinecarboxamide-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P264;P270;P271;P273;P280;P301+312;P302+352;P304+340;P305+351+338;P330;P337+313;P391;P403+233;
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3535
pictograms GHS07; GHS09
hstatements H302;H315;H319;H335;H411
Availability details for
Quantity
  • Availability
  • Add to Cart