ADV428293403 AChemBlock

diisopropyl 2-tosyl-2-azaspiro[3.3]heptane-6,6-dicarboxylate

  • MDL number MFCD27987092
  • Cas Number : 1237542-10-7
  • Molecular Weight : 423.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-azaspiro-3-3-heptane-6-6-dicarboxylic-acid-2-4-methylphenyl-sulfonyl-6-6-bis-1-methylethyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7767
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart