ADV428293412 AChemBlock

5,7-Dichloro-2-ethyl-2H-pyrazolo[4,3-D]pyrimidine

  • MDL number MFCD27987095
  • Cas Number : 1823331-38-9
  • Molecular Weight : 217.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-7-dichloro-2-ethyl-2h-pyrazolo-4-3-d-pyrimidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7776
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart