ADV428293415 AChemBlock

3-Amino-cyclobutanecarboxylic acid hydrochloride

  • MDL number MFCD19442672
  • Cas Number : 1201190-01-3
  • Molecular Weight : 151.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-amino-cyclobutanecarboxylic-acid-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7780
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart