ADV428293417 AChemBlock

tert-butyl N-(2-oxopiperidin-4-yl)carbamate

  • MDL number MFCD17170058
  • Cas Number : 1263281-78-2
  • Molecular Weight : 214.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-2-oxopiperidin-4-yl-carbamate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7782
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart