ADV428293423 AChemBlock

2-Azaspiro[3.3]heptane-6-carboxylic acid TFA salt

  • MDL number MFCD19982773
  • Cas Number : 1523572-02-2
  • Molecular Weight : 255.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form white solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-azaspiro-3-3-heptane-6-carboxylic-acid-tfa-salt.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3540
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart