ADV428293425 AChemBlock

3-Amino-3-methylazetidine dihydrochloride

  • MDL number MFCD09264377
  • Cas Number : 124668-47-9
  • Molecular Weight : 159.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-amino-3-methylazetidine-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3560
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart