ADV428293428 AChemBlock

2,3,5,6,8,9-hexahydro-1,4,7,10-Tetraoxacyclododecino[2,3-g]quinolin-15(12H)-one

  • MDL number MFCD22573388
  • Cas Number : 1355620-88-0
  • Molecular Weight : 291.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-3-5-6-8-9-hexahydro-1-4-7-10-tetraoxacyclododecino-2-3-g-quinolin-15-12h-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7794
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart