ADV428293439 AChemBlock

tert-butyl 6-formyl-2-azaspiro[3.3]heptane-2-carboxylate

  • MDL number MFCD24857302
  • Cas Number : 1440960-67-7
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form White solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-azaspiro-3-3-heptane-2-carboxylic-acid-6-formyl-1-1-dimethylethyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7805
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart