ADV428293455 AChemBlock

(1R,3S,4S)-3-(Boc-amino)-4-hydroxy-cyclohexanecarboxylic acid ethyl ester

  • MDL number MFCD23106246
  • Cas Number : 1210348-16-5
  • Molecular Weight : 287.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1r-3s-4s-3-boc-amino-4-hydroxy-cyclohexanecarboxylic-acid-ethyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7823
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart