ADV428293457 AChemBlock

Ethyl (1S,3R,4R)-4-{[(tert butoxy)carbonyl]amino}-3-hydroxycyclohexane-1-carboxylate

  • MDL number MFCD27987360
  • Cas Number : 1392745-47-9
  • Molecular Weight : 287.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1s-3r-4r-4-tert-butoxy-carbonyl-amino-3-hydroxycyclohexane-1-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7825
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart