ADV428293477 AChemBlock

tert-Butyl 5-(hydroxymethyl)thiazol-2-ylcarbamate

  • MDL number MFCD16877433
  • Cas Number : 1001419-37-9
  • Molecular Weight : 230.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-5-hydroxymethyl-thiazol-2-ylcarbamate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7846
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart