ADV428293501 AChemBlock

3-Bromo-2-methoxy-4-pyridinamine

  • MDL number MFCD11656225
  • Cas Number : 215364-86-6
  • Molecular Weight : 203.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-2-methoxy-4-pyridinamine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7871
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart