ADV428293510 AChemBlock

Diethyl 3-methyl-1H-pyrrole-2,4-dicarboxylate

  • MDL number MFCD00030385
  • Cas Number : 5448-16-8
  • Molecular Weight : 225.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form pale-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/diethyl-3-methyl-1h-pyrrole-2-4-dicarboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7880
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart