ADV428293513 AChemBlock

7-Amino-1,2,3,4-tetrahydro-2-methyl-7-Isoquinoline

  • MDL number MFCD11934487
  • Cas Number : 14097-40-6
  • Molecular Weight : 162.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-amino-1-2-3-4-tetrahydro-2-methyl-7-isoquinoline.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7883
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart