ADV428293518 AChemBlock

Ethyl 3-oxo-2-azaspiro[4.5]decane-8-carboxylate

  • MDL number MFCD20488669
  • Cas Number : 1073559-59-7
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-3-oxo-2-azaspiro-4-5-decane-8-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7888
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart