ADV428293519 AChemBlock

8-Amino-2-Boc-2-azaspiro[4.5]decane

  • MDL number MFCD22033481
  • Cas Number : 1363381-61-6
  • Molecular Weight : 254.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/8-amino-2-boc-2-azaspiro-4-5-decane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7889
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart