ADV428293529 AChemBlock

6-Bromo-2-oxa-spiro[3.3]heptane

  • MDL number MFCD22419347
  • Cas Number : 1363380-78-2
  • Molecular Weight : 177.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-2-oxa-spiro-3-3-heptane.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7899
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart