ADV428293532 AChemBlock

1-Boc-1,7-diaza-spiro[4.5]decane

  • MDL number MFCD11223544
  • Cas Number : 960294-14-8
  • Molecular Weight : 240.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-boc-1-7-diaza-spiro-4-5-decane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7902
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart