ADV428293534 AChemBlock

9-Amino-3-Boc-3-azaspiro[5.5]undecane

  • MDL number MFCD18711593
  • Cas Number : 1272758-41-4
  • Molecular Weight : 268.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/9-amino-3-boc-3-azaspiro-5-5-undecane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7904
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart