ADV428293537 AChemBlock

5-Chloropyrazolo[1,5-a]pyrimidine-3-carboxylic acid

  • MDL number MFCD20482240
  • Cas Number : 1315364-91-0
  • Molecular Weight : 197.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-chloropyrazolo-1-5-a-pyrimidine-3-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7907
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart