ADV428293546 AChemBlock

3-Fluoro-3-iodomethyloxetane

  • MDL number MFCD22566187
  • Cas Number : 1363381-23-0
  • Molecular Weight : 215.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-fluoro-3-iodomethyloxetane.html

Safety Information

signalword Danger
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7918
pictograms GHS06; GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart