ADV428293547 AChemBlock

1-(Aminomethyl)-3,3-difluoro-cyclobutanemethanol

  • MDL number MFCD22566199
  • Cas Number : 1363383-40-7
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-aminomethyl-3-3-difluoro-cyclobutanemethanol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7919
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart