ADV428293550 AChemBlock

1-Boc-3-fluoropiperidine-3-methanol

  • MDL number MFCD16250809
  • Cas Number : 1209781-11-2
  • Molecular Weight : 233.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-fluoropiperidine-3-methanol.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7922
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart