ADV428293553 AChemBlock

3-Methyl-1H-indazole-7-boronic acid

  • MDL number MFCD12028649
  • Cas Number : 1310404-47-7
  • Molecular Weight : 175.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-methyl-1h-indazole-7-boronic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7925
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart