ADV428293554 AChemBlock

1-Boc-2-(bromomethyl)azetidine

  • MDL number MFCD22566236
  • Cas Number : 1363380-84-0
  • Molecular Weight : 250.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Colorless/Light yellow liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-boc-2-bromomethyl-azetidine.html

Safety Information

signalword Danger
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7926
pictograms GHS05; GHS06; GHS07
hstatements H301;H312;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart