ADV428293559 AChemBlock

3-Hydroxymethyl-4-methylpyridine

  • MDL number MFCD01444898
  • Cas Number : 4664-27-1
  • Molecular Weight : 123.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-hydroxymethyl-4-methylpyridine.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7931
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart