ADV428293561 AChemBlock

3-(Chloromethyl)pyridazine hydrochloride

  • MDL number MFCD19705404
  • Cas Number : 27349-66-2
  • Molecular Weight : 165.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form brown solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-chloromethyl-pyridazine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2225
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart