ADV428293572 AChemBlock

2-Amino-6-cyanopyrazine

  • MDL number MFCD10697805
  • Cas Number : 59489-39-3
  • Molecular Weight : 120.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-amino-6-cyanopyrazine.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7944
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart