ADV428293579 AChemBlock

4-Chloro-2,3-dihydro-1H-pyrrolo[3,2-c]pyridine hydrochloride

  • MDL number MFCD14586358
  • Cas Number : 1241725-93-8
  • Molecular Weight : 191.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/4-chloro-2-3-dihydro-1h-pyrrolo-3-2-c-pyridine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/ac_sds?Catalog=G-7951
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart