ADV428293583 AChemBlock

cis-3-(Boc-amino)-4-methylpiperidine

  • MDL number MFCD18331628
  • Cas Number : 250275-20-8
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form light-yellow solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/cis-3-boc-amino-4-methylpiperidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7955
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart