ADV428293584 AChemBlock

Methyl 3-fluoroazetidine-3-carboxylate hydrochloride

  • MDL number MFCD27988017
  • Cas Number : 1421920-61-7
  • Molecular Weight : 169.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form White crystal
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-fluoroazetidine-3-carboxylate-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7956
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart