ADV428293587 AChemBlock

3-Chloro-4-methylpyridazine

  • MDL number MFCD08460979
  • Cas Number : 68206-04-2
  • Molecular Weight : 128.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-chloro-4-methylpyridazine.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7959
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart