ADV428293590 AChemBlock

6-Chloro-3-pyridazinecarboxaldehyde

  • MDL number MFCD10698041
  • Cas Number : 303085-53-2
  • Molecular Weight : 142.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Light yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-3-pyridazinecarboxaldehyde.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7962
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart