ADV428293596 AChemBlock

5-Amino-3-(difluoromethyl)-1H-pyrazole

  • MDL number MFCD15976291
  • Cas Number : 1284220-49-0
  • Molecular Weight : 133.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-amino-3-difluoromethyl-1h-pyrazole.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7968
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart