ADV428293598 AChemBlock

1-Boc-3,3-difluoro-4-oxopyrrolidine

  • MDL number MFCD16140377
  • Cas Number : 1215071-16-1
  • Molecular Weight : 221.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-3-difluoro-4-oxopyrrolidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7970
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart