ADV428293604 AChemBlock

(2R)-2-(Methoxymethyl)azetidine

  • MDL number MFCD11977227
  • Cas Number : 935668-82-9
  • Molecular Weight : 101.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light yellow solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2r-2-methoxymethyl-azetidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7976
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart