ADV428293609 AChemBlock

cis-(3-Aminocyclobutyl)acetic acid hydrochloride

  • MDL number MFCD27988022
  • Cas Number : 1523571-91-6
  • Molecular Weight : 165.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form White solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/cis-3-aminocyclobutyl-acetic-acid-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7981
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart