ADV428293622 AChemBlock

1-Boc-3-ethynyl-3-hydroxypyrrolidine

  • MDL number MFCD23106002
  • Cas Number : 275387-76-3
  • Molecular Weight : 211.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-ethynyl-3-hydroxypyrrolidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7994
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart