ADV428293623 AChemBlock

(S)-3,3-Difluorocyclopentanamine hydrochloride

  • MDL number MFCD23106013
  • Cas Number : 1408148-48-0
  • Molecular Weight : 157.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/s-3-3-difluorocyclopentanamine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7995
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart