ADV428293624 AChemBlock

3,3-Difluoro-cyclopentanemethanol

  • MDL number MFCD22199415
  • Cas Number : 883731-63-3
  • Molecular Weight : 136.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-3-difluoro-cyclopentanemethanol.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7996
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart