ADV428293638 AChemBlock

3-Methyltetrahydrofuran-3-carboxylic acid

  • MDL number MFCD12406628
  • Cas Number : 1158760-45-2
  • Molecular Weight : 130.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-methyltetrahydrofuran-3-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9010
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart