ADV428293648 AChemBlock

Methyl 2-Boc-2-aza-spiro[3.3]heptane-6-carboxylate

  • MDL number MFCD23106118
  • Cas Number : 1408074-81-6
  • Molecular Weight : 255.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-boc-2-aza-spiro-3-3-heptane-6-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9021
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart